Order Entry
Austria
ContactUsLinkComponent
Iron(III) chloride 27,5% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Corrosive
Irritant
Iron(III) chloride 27,5% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Catalog # 24207.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Iron(III) chloride 27,5% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Catalog # 24207.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    27.5 - 29.0 %
  • Acidity:
    Max. 0.08 meq/g
  • Density (20/4):
    1.260 - 1.280
  • Free chlorine:
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 30 ppm
  • Not precipitated by NH4OH (as SO4):
    Max. 0.05 %
  • NH4 (Ammonium):
    Max. 20 ppm
  • NO3 (Nitrate):
    Max. 20 ppm
  • PO4 (Phosphate):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • As (Arsenic):
    Max. 5 ppm
  • Fe (II) (Iron):
    Max. 30 ppm
  • Zn (Zinc):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
 :GHS05
 :GHS07
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005