Order Entry
Austria
ContactUsLinkComponent
Iron(III) chloride 27% in aqueous solution, GPR RECTAPUR®
Irritant
Corrosive
Iron(III) chloride 27% in aqueous solution, GPR RECTAPUR®
Catalog # 24206.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Iron(III) chloride 27% in aqueous solution, GPR RECTAPUR®
Catalog # 24206.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    27 - 29 %
  • Density (20/4):
    1.260 - 1.280
  • Heavy metals (as Pb):
    Max. 100 ppm
  • Not precipitated by NH4OH (as SO4):
    Max. 0.5 %
  • SO4 (Sulphate):
    Max. 0.02 %

Specification Test Results

About this item

 :
 :162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
 :GHS07
 :GHS05
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005