Order Entry
Austria
ContactUsLinkComponent
Iron(III) chloride (10% Fe) in aqueous solution, EMSURE® for analysis, Supelco®
Corrosive
Irritant
Iron(III) chloride (10% Fe) in aqueous solution, EMSURE® for analysis, Supelco®
Catalog # 1.05512.0250
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Iron(III) chloride (10% Fe) in aqueous solution, EMSURE® for analysis, Supelco®
Catalog # 1.05512.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.29 g/cm3 (20 °C)
  • pH:
     - 1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :162.21 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ300
EINECS:231-729-4
UN:2582
 :GHS05
 :GHS07
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005