Order Entry
Austria
ContactUsLinkComponent
Zinc chloride 71% ≥71% in aqueous solution
Zinc chloride 71% ≥71% in aqueous solution
  29142.290
 :  
undefined
Zinc chloride 71% ≥71% in aqueous solution
  29142.290
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

 

 

  • Assay:
    Min. 71 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm

 

 

 : Zinc dichloride, Dichlorozinc, Zinc (II) chloride
 :
 :136.3 g/mol
 :231-592-0
 :1840
 :GHS05
 :GHS07
 :GHS09
 :Ambient
 :7646-85-7
 :ZnCl₂
 :MFCD00011295
 :732
 :283
 :8 III
 :13,10185