

Ammonium sulphide 20% in aqueous solution, Sigma-Aldrich®
: Merck
:12135-76-1
:
:H₈N₂S
:MFCD00010892
:68.14 g/mol
:8,II
:M-Clarity™ quality level = MQ300
:235-223-4
:2683
:GHS02
:GHS05
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals