Order Entry
Austria
ContactUsLinkComponent
 
 
 
Ammonium sulphide 20% in aqueous solution, AnalaR NORMAPUR® analytical reagent
  21354.298
 :  
undefined
 
 
Ammonium sulphide 20% in aqueous solution, AnalaR NORMAPUR® analytical reagent
  21354.298
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay (as (NH4)2S from sulphide):
    17.0 - 23.0 %
  • Ignition residue (SO4):
    Max. 400 ppm
  • Cl (Chloride):
    Max. 0.02 %
  • CO3 (Carbonate):
    Max. 100 ppm

 

 

 :
 :68.14 g/mol
 :235-223-4
 :2683
 :8 (3, 6.1),II
 :GHS02
 :GHS05
 :12135-76-1
 :H₈N₂S
 :MFCD00010892