Order Entry
Austria
ContactUsLinkComponent
Ammonium sulphide 20% in aqueous solution analytical reagent, Supelco®
Flammable
Corrosive
Ammonium sulphide 20% in aqueous solution analytical reagent, Supelco®
Catalog # 1.05442.1000
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Ammonium sulphide 20% in aqueous solution analytical reagent, Supelco®
Catalog # 1.05442.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Vapor Pressure:
    450 hPa (20 °C)
  • Flash Point:
    15 °C
  • Explosion Limit:
    4 - 46 %(V)
  • Density:
    0.997 g/cm3 (20 °C)
  • pH:
    9.5 (200 g/l, H2O, 25 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

About this item

 :
 :68.14 g/mol
ADR:8 (3, 6.1),II
 :M-Clarity™ quality level = MQ100
EINECS:235-223-4
UN:2683
 :GHS02
 :GHS05
Storage Temperature:Ambient
CAS Number:12135-76-1
Formula:H₈N₂S
MDL Number:MFCD00010892