Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution 30 - 33%

Ammonia solution 30 - 33%

Corrosive
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Ammonia solution 30 - 33%
Supplier:  Roth Carl
Marketsource
Certificates
Ammonia solution 30 - 33%

2 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    200 mL
    Log in for availability
    Log in for availability
    1 * 200 mL
    1 * 200 mL - 22,40 €
    22,40 €
    1 * 200 mL
       22,40 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.