Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Ammonia solution ∼32%, GPR RECTAPUR®
 
undefined
 
 
 
Ammonia solution ∼32%, GPR RECTAPUR®
 

 

  • Assay:
    28 - 33 %
  • Density (20/4):
    0.880 - 0.910
  • Evaporation residue:
    Max. 100 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • CO3 (Carbonate):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 1 ppm

 

Frequently Bought Together