Order Entry
Austria
ContactUsLinkComponent
Ammonia solution 25% (NH₃ basis) in water, Puriss. p.a., Fluka™
Irritant
Corrosive
Environment
Ammonia solution 25% (NH₃ basis) in water, Puriss. p.a., Fluka™
Catalog # 09861-1L
CAS Number:  
Irritant
Corrosive
Environment
Ammonia solution 25% (NH₃ basis) in water, Puriss. p.a., Fluka™
Catalog # 09861-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (acidimetric):
    23.0 - 27.0 %
  • Non-volatile matter:
    Max. 0.0018 %
  • Density (D 20/4):
    0.890 - 0.915
  • Silver (Ag):
    Max. 0.02 ppm
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.05 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 1 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.05 ppm
  • Chromium (Cr):
    Max. 0.05 ppm
  • Copper (Cu):
    Max. 0.1 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.02 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.05 ppm
  • Molybdenum (Mo):
    Max. 0.05 ppm
  • Sodium (Na):
    Max. 1 ppm
  • Nickel (Ni):
    Max. 0.05 ppm
  • Lead (Pb):
    Max. 0.05 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Thallium (Tl):
    Max. 0.05 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • Chloride (Cl):
    Max. 0.5 ppm
  • Phosphate (PO₄):
    Max. 0.5 ppm
  • Sulfate (SO₄):
    Max. 2 ppm
  • Sulfide (S):
    Max. 0.2 ppm
  • Carbonate (as CO₂):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:37.7 °C (1.013 hPa)
Melting Pt:–92 °C
Density:0.90 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :Packed in coated, shock-and leak-protected glass bottle
 :GHS07
 :GHS05
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650