Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution 25%, TECHNICAL

Ammonia solution 25%, TECHNICAL

Corrosive
Irritant
Environment
Supplier:   VWR Chemicals
Ammonia solution 25%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Ammonia solution 25%, TECHNICAL

2 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.907 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 24 - 26 %
Product Family Options

Product Information