


Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
ADR:8,III
:M-Clarity™ quality level = MQ300
UN:2672
:GHS05
:GHS07
:GHS09
Specification Test Results
- Assay (acidimetric, NH₃):25.0 - 30.0 %
- Identity:passes test
- Appearance of solution:passes test
- Density (20/20°C):0.892 - 0.910
- Carbonate (as CO₂):≤ 10 ppm
- Chloride (Cl):≤ 0.5 ppm
- Phosphate (PO₄):≤ 0.5 ppm
- Silicon (as SiO₂):≤ 10 ppm
- Sulfate (SO₄):≤ 2 ppm
- Sulphide (S):≤ 0.2 ppm
- Pyridine and related substances:≤ 2 ppm
- Ag (Silver):≤ 0.020 ppm
- Al (Aluminium):≤ 0.500 ppm
- Au (Gold):≤ 0.100 ppm
- Ba (Barium):≤ 0.050 ppm
- Bi (Bismuth):≤ 0.100 ppm
- Ca (Calcium):≤ 0.500 ppm
- Cd (Cadmium):≤ 0.050 ppm
- Co (Cobalt):≤ 0.050 ppm
- Cr (Chromium):≤ 0.050 ppm
- Cu (Copper):≤ 0.100 ppm
- Fe (Iron):≤ 0.100 ppm
- Ga (Gallium):≤ 0.020 ppm
- In (Indium):≤ 0.020 ppm
- K (Potassium):≤ 0.500 ppm
- Li (Lithium):≤ 0.020 ppm
- Mg (Magnesium):≤ 0.100 ppm
- Mn (Manganese):≤ 0.050 ppm
- Mo (Molybdenum):≤ 0.050 ppm
- Na (Sodium):≤ 0.500 ppm
- Ni (Nickel):≤ 0.050 ppm
- Pb (Lead):≤ 0.050 ppm
- Pt (Platinum):≤ 0.100 ppm
- Sn (Tin):≤ 0.100 ppm
- Sr (Strontium):≤ 0.100 ppm
- Ti (Titanium):≤ 0.100 ppm
- Tl (Thallium):≤ 0.050 ppm
- Zn (Zinc):≤ 0.100 ppm
- Oxidizable substances:passes test
- Substances reducing potassium permanganate (as O):≤ 5 ppm
- Residue on ignition (as SO₄):≤ 10 ppm
- Non volatile matter:≤ 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.05432.1000_TDS_EN ProductPage_105432
Produktspezifikation
Merck Pharma Segment Brochure
Produktliteratur
msds_105432
Sicherheitsdatenblatt (SDS)
Frequently Bought Together