Ammonia solution 25% in water, extra pure
: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
:1336-21-6
:7664-41-7
:NH₃
:MFCD00011418
:32…37.7 °C (1013 hPa)
:–57 °C
:0.91 g/cm³
:35.05 g/mol
:215-647-6
:2672
:8 III
:GHS07
:GHS05
:GHS09
:Refrigerator
- Environmentally Preferable:
- Appearance:Clear colorless Solution
- Titration with HCl:≥25 % (As NH3)
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤50 ppm
Frequently Bought Together