

Ammoniaklösung 20%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Density:0,92 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay:18 - 22 %
- Density (20/4):0.910 - 0.930
- Evaporation residue:Max. 0.02 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 20 ppm
- Fe (Iron):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
spec_VWRC21180.242 spec_VWRC21180.242 spec_VWRC21180.297 spec_VWRC21180.297 spec_VWRC21180.366
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together