



Tetramethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Melting Pt:–98 °C
Density:0.9 g/cm³
Flash Pt:6 °C
EINECS:200-882-9
UN:3286
ADR:3,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Titration with HCl:24.5 to 26.0 %
- HPLC:≥97.5 % (ELSD)
- Stabilizer:2 % CaCO3
- Specific optical rotation:+185° to +195° (20°C, 589 nm) (c=1, ethyl acetate)
Specification Test Results
Frequently Bought Together