

Tetramethylammonium hydroxide 25% in aqueous solution
MOLEM26136897
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:91.15 g/mol
:110 °C (1013 hPa)
:1.02 g/cm³ (20 °C)
:Ambient
:200-882-9
:1835
:8,II
:GHS06
:GHS05
:75-59-2
:C₄H₁₃NO
:MFCD00008280