Order Entry
Austria
ContactUsLinkComponent
Tetramethylammonium hydroxide 25% in aqueous solution
Toxic
Corrosive
Tetramethylammonium hydroxide 25% in aqueous solution
Catalog # HONE17741.5000
CAS Number:  
Toxic
Corrosive
Tetramethylammonium hydroxide 25% in aqueous solution
Catalog # HONE17741.5000
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-882-9
UN:1835
ADR:8,II
 :GHS06
 :GHS05
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280