Tetramethylammonium hydroxide 10% in aqueous solution, Sigma-Aldrich®
: Merck
:75-59-2
:
:C₄H₁₃NO
:MFCD00008280
:91.15 g/mol
:110 °C (1013 hPa)
:1.02 g/cm³ (20 °C)
:200-882-9
:1835
:8,II
:M-Clarity™ quality level = MQ100
:GHS06
:GHS05
:Ambient
Frequently Bought Together