Order Entry
Austria
ContactUsLinkComponent
 
 
Sodium hydroxide 25% in aqueous solution, J.T.Baker®
  2613.9950
 :  Avantor
 :  
 
Sodium hydroxide 25% in aqueous solution, J.T.Baker®
  2613.9950
 :  Avantor
 :  

 

  • Pk:
    950 L
  • Seq:
    5783

 

 

  • Assay (NaOH):
    24.4 - 25.6 %
  • Appearance (clear, colorless liquid):
    passes test
  • Endotoxin Concentration (EU/ml):
    max. 2.0
  • Sodium Carbonate (Na2CO3):
    max. 0.6 %
  • Trace Impurities (in ppm)Chlorides (as Cl):
    max. 5
  • Trace Impurities (in ppm)Iron (Fe):
    max. 0.5
  • Trace Impurities (in ppm)Heavy Metals (as Pb):
    max. 1

 

 

 :
 :40 g/mol
 :119…135 °C (1013 hPa)
 :9…10 °C
 :1,53 g/cm³ (20 °C)
 :Ambient
 :215-185-5
 :1824
 :8,II
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548