Potassium hydroxide 50% in aqueous solution, Selectipur® for the electronics industry
Catalog # 56999579.
Supplier: BASF ELECTRONIC CHEMICALS
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:300 kg
- Seq:4792
Specifications
About this item
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553