Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 47% in aqueous solution, EMSURE® for analysis, Supelco®
Corrosive
Irritant
Potassium hydroxide 47% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Corrosive
Irritant
Potassium hydroxide 47% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.475 g/cm3 (20 °C)
  • pH:
    13.5 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together