Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 45.0 %
  • Ammonium Hydroxide Precipitate:
    max. 0.005 %
  • Chloride (Cl):
    max. 0.005 %
  • Potassium Carbonate (K2CO3):
    max. 0.5 %
  • Sodium (Na) (by AAS):
    max. 0.02 %
  • Sulfate (SO4):
    max. 0.001 %
  • Iron (Fe):
    max. 2
  • Nickel (Ni):
    max. 5
  • Heavy Metals (as Ag):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5
  • Phosphate (PO4):
    max. 2

Specification Test Results

Related Information

Frequently Bought Together