
Sulphuric acid 93-98%, Environmental Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,8974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
Related Information
Frequently Bought Together