Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 62% analytical reagent, for the determination of fats in cheese, Supelco®
Sulphuric acid 62% analytical reagent, for the determination of fats in cheese, Supelco®
Supplier:  Merck
Certificates
Sulphuric acid 62% analytical reagent, for the determination of fats in cheese, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Density:
    1.52 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly acid

Specification Test Results

Related Information

Frequently Bought Together