Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 40%, Supelco®
Corrosive
Sulphuric acid 40%, Supelco®
Catalog # 1.09286.2500
Supplier:  Merck
CAS Number:  
Corrosive
Sulphuric acid 40%, Supelco®
Catalog # 1.09286.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Melting Point:
    -68 °C
  • Boiling Point:
    119 °C
  • Density:
    1.30 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly acid

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1.1…1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
 :M-Clarity™ quality level = MQ300
EINECS:231-639-5
UN:2796
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589