Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 37%, Supelco®
Corrosive
Sulphuric acid 37%, Supelco®
Catalog # 4.80104.9008
Supplier:  Merck
CAS Number:  
Corrosive
Sulphuric acid 37%, Supelco®
Catalog # 4.80104.9008
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic jerrican
  • Pk:
    8 L
  • Seq:
    5210

Specifications

About this item

 :
 :98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1.1…1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:231-639-5
UN:2796
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589