Silver sulphate 10 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
EINECS:233-653-7
UN:1830
ADR:8,II
:GHS05
Specification Test Results
- Chemical oxygen demand (COD):Passes test
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250)
Packaging Information
27327353.pdf 27798953.pdf spec_VWRC30491.247 spec_VWRC30491.420
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together