Silver sulphate 10 g/l in sulphuric acid for COD determination, Supelco®
: Merck
:10294-26-5
:
:Ag₂SO₄
:MFCD00003407
:311.8 g/mol
:8,II
:M-Clarity™ quality level = MQ100
:233-653-7
:3264
:GHS05
:GHS09
:Ambient
- Solubility:(20 °C) soluble,(caution ! development of heat)
- Density:1.84 g/cm3 (20 °C)
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:8
- Hazard Class Description:Corrosive material
Frequently Bought Together