Order Entry
Austria
ContactUsLinkComponent
Silver sulphate 10 g/l in sulphuric acid volumetric solution, for COD determination according to DIN 38409-41, Fluka™
Corrosive
Silver sulphate 10 g/l in sulphuric acid volumetric solution, for COD determination according to DIN 38409-41, Fluka™
Certificates
undefined
Corrosive
Silver sulphate 10 g/l in sulphuric acid volumetric solution, for COD determination according to DIN 38409-41, Fluka™
Certificates

Specification Test Results

  • Assay of H₂SO₄:
    94 - 96 %
  • Assay of Ag₂SO₄:
    0.54 - 0.56 %
  • Density (20°C):
    1.83 - 1.85 g/ml

Specification Test Results

Related Information

Frequently Bought Together