

Silver nitrate 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.09080.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:169.87 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:231-853-9
UN:1760
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414