Order Entry
Austria
ContactUsLinkComponent
Nitric acid 69%, ARISTAR® for trace analysis
Oxidizing
Corrosive
Toxic
Nitric acid 69%, ARISTAR® for trace analysis
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Corrosive
Toxic
Nitric acid 69%, ARISTAR® for trace analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    68.5 - 69.5 %
  • Cl (Chloride):
    Max. 0.05 ppm
  • PO4 (Phosphate):
    Max. 0.1 ppm
  • Si (as silicate):
    Max. 0.1 ppm
  • SO4 (Sulphate):
    Max. 0.5 ppm
  • As + Sb (as As):
    Max. 0.005 ppm
  • Ag (Silver):
    Max. 0.005 ppm
  • Al (Aluminium):
    Max. 0.020 ppm
  • Au (Gold):
    Max. 0.005 ppm
  • B (Boron):
    Max. 0.005 ppm
  • Ba (Barium):
    Max. 0.005 ppm
  • Be (Beryllium):
    Max. 0.005 ppm
  • Bi (Bismuth):
    Max. 0.005 ppm
  • Ca (Calcium):
    Max. 0.05 ppm
  • Cd (Cadmium):
    Max. 0.005 ppm
  • Co (Cobalt):
    Max. 0.005 ppm
  • Cr (Chromium):
    Max. 0.010 ppm
  • Cu (Copper):
    Max. 0.005 ppm
  • Fe (Iron):
    Max. 0.020 ppm
  • Ga (Gallium):
    Max. 0.020 ppm
  • Ge (Germanium):
    Max. 0.010 ppm
  • Hg (Mercury):
    Max. 0.001 ppm
  • In (Indium):
    Max. 0.010 ppm
  • K (Potassium):
    Max. 0.010 ppm
  • Li (Lithium):
    Max. 0.005 ppm
  • Mg (Magnesium):
    Max. 0.010 ppm
  • Mn (Manganese):
    Max. 0.005 ppm
  • Mo (Molybdenum):
    Max. 0.005 ppm
  • Na (Sodium):
    Max. 0.050 ppm
  • Ni (Nickel):
    Max. 0.005 ppm
  • Pb (Lead):
    Max. 0.005 ppm
  • Pt (Platinum):
    Max. 0.005 ppm
  • Sn (Tin):
    Max. 0.020 ppm
  • Sr (Strontium):
    Max. 0.005 ppm
  • Ti (Titanium):
    Max. 0.005 ppm
  • Tl (Thallium):
    Max. 0.005 ppm
  • V (Vanadium):
    Max. 0.005 ppm
  • Zn (Zinc):
    Max. 0.010 ppm
  • Zr (Zirconium):
    Max. 0.005 ppm
  • Ignition residue (SO4):
    Max. 1 ppm

Specification Test Results

Frequently Bought Together