Order Entry
Austria
ContactUsLinkComponent
Silver nitrate 0.02 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
Silver nitrate 0.02 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 191265J
Supplier:  VWR Chemicals
CAS Number:  
undefined
Silver nitrate 0.02 mol/l (0.02 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 191265J
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0198 - 0.0202 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :169.87 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-853-9
UN:3082
ADR:9,III
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414