Iodine 0.5 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
1.09098.1000
: Merck
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
:
:253.81 g/mol
:8,II
:M-Clarity™ quality level = MQ100
:231-442-4
:3264
:GHS08
:Ambient
:7553-56-2
:I₂
:MFCD00011355