Iodine standard solution 0.05 mol/l (0.1 N) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Density:1.02 g/cm³
Storage Temperature:Ambient
EINECS:231-442-4
ADR:8,II
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark brown Solution
- Titration with Na2S2O3:0.0998 to 0.1002 N (20°C)
- CO2 absorption capacity:≥19.0 %
Specification Test Results
Frequently Bought Together