Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31960.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31960.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428