ContactUsLinkComponent
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Ammonium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • NIST traceable:
    Confirmed