Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Ammonium thiocyanate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4650.
Supplier:  Avantor
CAS Number:  
Ammonium thiocyanate concentrate for dilution to 0.1 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4650.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0998-0.1002

Specification Test Results

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428