Magnesium chloride 1.0 M MgCl2 volumetric solution, Fluka™
Catalog # 63020-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:0.995 - 1.005 mol/l
- Density (D 20/4):1.070 - 1.090
Specification Test Results
About this item
:
:95.21 g/mol
Merck Index:13,05684
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.08 g/cm³
Flash Pt:Does not flash
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106
Flash Pt: Does not flash