Magnesium chloride 1 mol/l (2 N) in aqueous solution
Catalog # ROCKMB-048
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
:95.21 g/mol
Merck Index:13,05684
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106