Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution for molecular biology
Catalog # A5324.0001
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
- Environmentally Preferable:
Specifications
About this item
:
:95.21 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106