ContactUsLinkComponent
Calcium chloride 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution
Calcium chloride 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution
  190464K
 :  
undefined
Calcium chloride 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution
  190464K
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • NIST traceable:
    Confirmed
 :
 :110.98 g/mol
 :233-140-8
 :GHS07
 :Ambient
 :10043-52-4
 :CaCl₂
 :MFCD00010903