Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, VOLUSOL® volumetric solution
Corrosive
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, VOLUSOL® volumetric solution
Catalog # 306452.2500
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, VOLUSOL® volumetric solution
Catalog # 306452.2500
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    3.992 - 4.008 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:<1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548