Order Entry
Austria
ContactUsLinkComponent
 
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, AVS TITRINORM volumetric solution low in carbonate
 
undefined
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, AVS TITRINORM volumetric solution low in carbonate
 

 

  • Environmentally Preferable:

 

 

  • Titer (20°C; real value 0.2 % accuracy):
    3.992 - 4.008 mol/l
  • CO₃ (as Na₂CO₃):
    Max. 0.2 %
  • NIST traceable:
    Confirmed

 

Frequently Bought Together