
Sodium hydroxide 2 mol/l (2 N) in aqueous solution
Supplier: Roth Carl
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Frequently Bought Together
Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck