Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution 0.5 M NaOH (0.5N) volumetric solution, Fluka™
: Caustic soda
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:>100 °C (1.013 hPa)
:1.020 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,III
:GHS05
- Environmentally Preferable:
- Assay:0.4900 - 0.5100 N
- Carbonate (as Na₂CO₃):Max. 0.010 N
- Traceable to NIST SRM:complying
- Vapor Density:>1 (vs air)
Frequently Bought Together