Potassium hydroxide 8 mol/l (8 N) in water, pure
:1310-58-3
:
:KOH
:MFCD00003553
:56.11 g/mol
:1.326 g/cm³
:Ambient
:215-181-3
:1814
:8,II
:GHS05
:GHS07
- Appearance:Clear colorless Solution
- Strength:7.984 to 8.016 N (20°C)
- Composition:448 g/l (g KOH/l H20)
Frequently Bought Together