Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 5 mol/l (5 N) in aqueous solution
Potassium hydroxide 5 mol/l (5 N) in aqueous solution
Catalog # MC310476.1000
Supplier:  PROLABO CMP
CAS Number:  
Potassium hydroxide 5 mol/l (5 N) in aqueous solution
Catalog # MC310476.1000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553