Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution (max. 0.00004% Ca), Supelco®
Potassium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution (max. 0.00004% Ca), Supelco®
Catalog # 1.09107.1000
Supplier:  Merck
CAS Number:  
Potassium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® volumetric solution (max. 0.00004% Ca), Supelco®
Catalog # 1.09107.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:1814
 :GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553