ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.499 - 0.501 mol/l
  • NIST traceable:
    Confirmed