
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
Catalog # ROTHN071.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
About this item
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553